Merck
  • 2-substituted 3-(aminooxy)propanamines as inhibitors of ornithine decarboxylase: synthesis and biological activity.
2-substituted 3-(aminooxy)propanamines as inhibitors of ornithine decarboxylase: synthesis and biological activity.
Journal of medicinal chemistry (1992-04-17)
J Stanek, J Frei, H Mett, P Schneider, U Regenass
ABSTRACT
1-Amino-3-(aminooxy)-2-propanol (6a) has been synthesized and found to inhibit rat liver ornithine decarboxylase (ODC) with an IC50 in the nanomolar range. Compound 6a served as a basis for the design of new enzyme inhibitors, which led to the identification of 3-(aminooxy)-2-fluoropropanamine (15) as a new powerful enzyme blocker. Compound 15 inhibited ODC at 3 times lower concentrations than 6a and 3-(aminooxy)propanamine (APA), and it was superior to APA as an antiproliferative agent in inhibiting the growth of human T24 bladder carcinoma cells in vitro.
Sigma-Aldrich
Ethyl acetohydroxamate, 97%