Merck
  • Macrophage ABHD5 promotes colorectal cancer growth by suppressing spermidine production by SRM.
Macrophage ABHD5 promotes colorectal cancer growth by suppressing spermidine production by SRM.
Nature communications (2016-05-18)
Hongming Miao, Juanjuan Ou, Yuan Peng, Xuan Zhang, Yujuan Chen, Lijun Hao, Ganfeng Xie, Zhe Wang, Xueli Pang, Zhihua Ruan, Jianjun Li, Liqing Yu, Bingzhong Xue, Hang Shi, Chunmeng Shi, Houjie Liang
ABSTRACT
Metabolic reprogramming in stromal cells plays an essential role in regulating tumour growth. The metabolic activities of tumour-associated macrophages (TAMs) in colorectal cancer (CRC) are incompletely characterized. Here, we identify TAM-derived factors and their roles in the development of CRC. We demonstrate that ABHD5, a lipolytic co-activator, is ectopically expressed in CRC-associated macrophages. We demonstrate in vitro and in mouse models that macrophage ABHD5 potentiates growth of CRC cells. Mechanistically, ABHD5 suppresses spermidine synthase (SRM)-dependent spermidine production in macrophages by inhibiting the reactive oxygen species-dependent expression of C/EBPɛ, which activates transcription of the srm gene. Notably, macrophage-specific ABHD5 transgene-induced CRC growth in mice can be prevented by an additional SRM transgene in macrophages. Altogether, our results show that the lipolytic factor ABHD5 suppresses SRM-dependent spermidine production in TAMs and potentiates the growth of CRC. The ABHD5/SRM/spermidine axis in TAMs might represent a potential target for therapy.
Product Description
Roche
Collagenase B, from Clostridium histolyticum
Pricing and availability is not currently available.
Millipore
Thioglycollate medium, NutriSelect® Plus, USP, EP, AOAC®, powder, pkg of 500 g, pkg of 1000 g, for bacteria
Pricing and availability is not currently available.