- Unsafe SETs: histone lysine methyltransferases and cancer.
Unsafe SETs: histone lysine methyltransferases and cancer.
Trends in biochemical sciences (2002-08-02)
Robert Schneider, Andrew J Bannister, Tony Kouzarides
PMID12151224
ABSTRACT
Enzymes that covalently modify histones control many cellular processes by affecting gene expression. A new class of these enzymes is the histone lysine methyltransferase family, whose catalytic activity lies within a conserved domain, the SET domain. This article surveys the evidence for a connection between SET-domain-containing proteins and cancer. It proposes that deregulation of SET-domain function has an important role in carcinogenesis.