Merck
  • Copper-catalyzed amine-alkyne-alkyne addition reaction: an efficient method for the synthesis of gamma,delta-alkynyl-beta-amino acid derivatives.
Copper-catalyzed amine-alkyne-alkyne addition reaction: an efficient method for the synthesis of gamma,delta-alkynyl-beta-amino acid derivatives.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-09-25)
Lei Zhou, Qi Shuai, Huan-feng Jiang, Chao-Jun Li
ABSTRACT
A simple and efficient method for the synthesis of gamma,delta-alkynyl-beta-amino acid derivatives by a copper-catalyzed three-component amine-alkyne-alkyne addition reaction was developed. Various gamma,delta-alkynyl-beta-amino acid derivatives were synthesized in moderate to good yields in one step. With chiral prolinol derivatives employed as the amine component, excellent diastereoselectivities (up to >99:1 diastereomeric ratio (dr)) were obtained. The scope of the reaction and further transformations of the resulting amino acid derivatives, such as deprotection and cyclization are also described.
Sigma-Aldrich
1-Ethynyl-3-fluorobenzene, 98%
Pricing and availability is not currently available.