Merck
  • Fast and sensitive LC-MS/MS method measuring human mycotoxin exposure using biomarkers in urine.
Fast and sensitive LC-MS/MS method measuring human mycotoxin exposure using biomarkers in urine.
Archives of toxicology (2014-09-12)
B Huybrechts, J C Martins, Ph Debongnie, S Uhlig, A Callebaut
ABSTRACT
A direct, fast and sensitive LC-MS/MS method was developed to measure biomarkers for mycotoxin exposure in human urine. In total, 32 biomarkers were quantitatively or semi-quantitatively measured in 32 urine samples of Belgian volunteers using two injections. All urine samples contained deoxynivalenol-15-glucuronide, the major detoxification metabolite of deoxynivalenol, in the ng/mL range. Also deoxynivalenol-3-glucuronide and de-epoxy-deoxynivalenol-glucuronide were present in, respectively, 90 and 25% of the samples, while deoxynivalenol was detected in 60% of the samples, in lower concentrations. Deoxynivalenol glucuronides were the major biomarkers for deoxynivalenol exposure. Ochratoxin A was detected in 70% of the samples in pg/mL. Citrinin and/or dihydrocitrinone were detected in 90% of the samples, also in concentrations of pg/mL. The presence of ochratoxin A and citrinin was confirmed by a second method using sample cleanup by immunoaffinity columns, followed by LC-MS/MS. Our data show that humans are much more exposed to citrinin than realized before and suggest further work on citrinin exposure in relation with ochratoxin A exposure, as both mycotoxins are nephrotoxic.
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
3-Acetyldeoxynivalenol, from Fusarium roseum
Sigma-Aldrich
15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum