Merck
  • The effect of RE substitution in layered REO(0.5)F(0.5)BiS2: chemical pressure, local disorder and superconductivity.
The effect of RE substitution in layered REO(0.5)F(0.5)BiS2: chemical pressure, local disorder and superconductivity.
Physical chemistry chemical physics : PCCP (2015-08-04)
Y Mizuguchi, E Paris, T Sugimoto, A Iadecola, J Kajitani, O Miura, T Mizokawa, N L Saini
ABSTRACT
We have studied the effect of RE substitution on the structure and the local atomic disorder in REO0.5F0.5BiS2 (RE = rare-earth) to understand their correlation with the bulk superconductivity in these materials. The mean RE size, affecting the chemical pressure, has been varied in two series namely Ce1-xNdxO0.5F0.5BiS2 and Nd1-ySmyO0.5F0.5BiS2. The lattice parameters evolve anomalously, showing an anisotropic shrinkage (elongation) of the c-axis (a-axis) to an isotropic expansion of the lattice with increasing mean RE size. The Bi L3-edge extended X-ray absorption fine structure (EXAFS) measurements are performed to investigate local displacements in the BiS2 lattice, revealing a large disorder and a sharp boundary between the Ce-containing and Sm-containing series with a distinct local structure. The results suggest that the bulk superconductivity in REO0.5F0.5BiS2 is correlated with anomalous atomic displacements in the Bi-S1 network, likely to be a combined effect of active Bi 6s electronic states and a possible Jahn-Teller-like instability of the Bi 6px/6py electrons.
Sigma-Aldrich
Boron nitride, powder, ~1 μm, 98%
Sigma-Aldrich
Boron nitride, nanopowder, <150 nm avg. part. size (TEM), 99% trace metals basis