Merck
  • Epigenetic Silencing of SPINT2 Promotes Cancer Cell Motility via HGF-MET Pathway Activation in Melanoma.
Epigenetic Silencing of SPINT2 Promotes Cancer Cell Motility via HGF-MET Pathway Activation in Melanoma.
The Journal of investigative dermatology (2015-04-25)
Soonyean Hwang, Hye-Eun Kim, Michelle Min, Rekha Raghunathan, Izabela P Panova, Ruchi Munshi, Byungwoo Ryu
ABSTRACT
Aberrant HGF-MET (hepatocyte growth factor-met proto-oncogene) signaling activation via interactions with surrounding stromal cells in tumor microenvironment has significant roles in malignant tumor progression. However, extracellular proteolytic regulation of HGF activation, which is influenced by the tumor microenvironment, and its consequential effects on melanoma malignancy remain uncharacterized. In this study, we identified SPINT2 (serine peptidase inhibitor Kunitz type 2), a proteolytic inhibitor of hepatocyte growth factor activator (HGFA), which has a significant role in the suppression of the HGF-MET pathway and malignant melanoma progression. SPINT2 expression is significantly lower in metastatic melanoma tissues compared with those in early-stage primary melanomas, which also corresponded with DNA methylation levels isolated from tissue samples. Treatment with the DNA-hypomethylating agent decitabine in cultured melanoma cells induced transcriptional reactivation of SPINT2, suggesting that this gene is epigenetically silenced in malignant melanomas. Furthermore, we show that ectopically expressed SPINT2 in melanoma cells inhibits the HGF-induced MET-AKT (v-Akt murine thymoma viral oncogene) signaling pathway and decreases malignant phenotype potential such as cell motility and invasive growth of melanoma cells. These results suggest that SPINT2 is associated with tumor-suppressive functions in melanoma by inhibiting an extracellular signal regulator of HGF, which is typically activated by tumor-stromal interactions. These findings indicate that epigenetic impairment of the tightly regulated cytokine-receptor communications in tumor microenvironment may contribute to malignant tumor progression.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Spint2
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human SPINT2