Merck
  • Type I phosphatidylinositol phosphate kinase beta regulates focal adhesion disassembly by promoting beta1 integrin endocytosis.
Type I phosphatidylinositol phosphate kinase beta regulates focal adhesion disassembly by promoting beta1 integrin endocytosis.
Molecular and cellular biology (2010-07-14)
Wei-Ting Chao, Felicity Ashcroft, Alexes C Daquinag, Tegy Vadakkan, Zhubo Wei, Pumin Zhang, Mary E Dickinson, Jeannette Kunz
ABSTRACT
Cell migration requires the regulated disassembly of focal adhesions, but the underlying mechanisms remain poorly defined. We have previously shown that focal adhesion disassembly requires the dynamin 2- and clathrin-dependent endocytosis of ligand-activated beta1 integrins. Here, we identify type I phosphatidylinositol phosphate kinase beta (PIPKIbeta), an enzyme that generates phosphatidylinositol-4,5-bisphosphate (PI4,5P(2)), as a key regulator of this process. We found that knockdown of PIPKIbeta by RNA interference blocks the internalization of active beta1 integrins and impairs focal adhesion turnover and cell migration. These defects are caused by the failure to target the endocytic machinery, including clathrin adaptors and dynamin 2, to focal adhesion sites. As a consequence, depletion of PIPKIbeta blocks clathrin assembly at adhesion plaques and prevents complex formation between dynamin 2 and focal adhesion kinase (FAK), a critical step in focal adhesion turnover. Together, our findings identify PIPKIbeta as a novel regulator of focal adhesion disassembly and suggest that PIPKIbeta spatially regulates integrin endocytosis at adhesion sites to control cell migration.