Merck
  • Dominant-negative alpha-subunit of farnesyl- and geranyltransferase inhibits glucose-stimulated, but not KCl-stimulated, insulin secretion in INS 832/13 cells.
Dominant-negative alpha-subunit of farnesyl- and geranyltransferase inhibits glucose-stimulated, but not KCl-stimulated, insulin secretion in INS 832/13 cells.
Diabetes (2006-12-29)
Rajakrishnan Veluthakal, Hitchintan Kaur, Marc Goalstone, Anjaneyulu Kowluru
ABSTRACT
The majority of small G-proteins undergo posttranslational modifications (e.g., isoprenylation) at their C-terminal cysteine residues. Such modifications increase their hydrophobicity, culminating in translocation of the modified proteins to their relevant membranous sites for interaction with their respective effectors. Previously, we reported glucose-dependent activation and membrane association of Rac1 in INS 832/13 cells. We also demonstrated modulatory roles for Rac1/GDP dissociation inhibitor in glucose-stimulated insulin secretion (GSIS) in INS 832/13 cells, further affirming roles for Rac1 in GSIS. Herein, we demonstrate that geranylgeranyltransferase inhibitor-2147 (GGTI-2147), an inhibitor of protein prenylation, markedly increased cytosolic accumulation of Rac1 and elicited significant inhibition of GSIS from INS 832/13 cells. In the current study, we also examined the localization of protein prenyltransferases (PPTases) and regulation of GSIS by PPTases in INS 832/13 cells. Western blot analyses indicated that the regulatory alpha-subunit and the structural beta-subunit of PPTase holoenzyme are predominantly cytosolic in their distribution. Overexpression of an inactive mutant of the regulatory alpha-subunit of PPTase markedly attenuated glucose- but not KCl-induced insulin secretion from INS 832/13 cells. Together, our findings provide the first evidence for the regulation of GSIS by PPTase in INS 832/13 cells. Furthermore, they support our original hypothesis that prenylation of specific G-proteins may be necessary for GSIS.