Merck
  • Prothrombin complex concentrates and a specific antidote to dabigatran are effective ex-vivo in reversing the effects of dabigatran in an anticoagulation/liver trauma experimental model.
Prothrombin complex concentrates and a specific antidote to dabigatran are effective ex-vivo in reversing the effects of dabigatran in an anticoagulation/liver trauma experimental model.
Critical care (London, England) (2014-02-07)
Oliver Grottke, Joanne van Ryn, Henri M H Spronk, Rolf Rossaint
ABSTRACT
New oral anticoagulants are effective alternatives to warfarin. However, no specific reversal agents are available for life-threatening bleeding or emergency surgery. Using a porcine model of trauma, this study assessed the ability of prothrombin complex concentrate (PCC), activated PCC (aPCC), recombinant FVIIa (rFVIIa) and a specific antidote to dabigatran (aDabi-Fab) to reverse the anticoagulant effects of dabigatran. Dabigatran etexilate (DE) was given orally for 3 days (30 mg/kg bid) and intravenously on day 4 to achieve consistent, supratherapeutic concentrations of dabigatran. Blood samples were collected at baseline, after oral DE, after intravenous dabigatran, and 60 minutes post-injury. PCC (30 and 60 U/kg), aPCC (30 and 60 U/kg), rFVIIa (90 and 180 μg/kg) and antidote (60 and 120 mg/kg) were added to blood samples ex-vivo. Coagulation was assessed by thromboelastometry, global coagulation assays and diluted thrombin time. Plasma concentrations of dabigatran were 380 ± 106 ng/ml and 1423 ± 432 ng/ml after oral and intravenous administration, respectively, and all coagulation parameters were affected by dabigatran. Both PCCs and aDabi-Fab, but not rFVIIa, reversed the effects of dabigatran on thromboelastometry parameters and prothrombin time. In contrast, aPTT was only normalised by aDabi-Fab. Plasma concentration (activity) of dabigatran remained elevated after PCC and rFVIIa therapy, but was not measureable after aDabi-Fab. In conclusion, PCC and aPCC were effective in reducing the anticoagulant effects of dabigatran under different conditions, while aDabi-Fab fully corrected all coagulation measures and decreased the plasma concentration of dabigatran below the limit of detection. No significant effects were observed with rFVIIa.