Merck
  • Synthesis and biological evaluation of (18)F-labeled Fluoroethoxy tryptophan analogues as potential PET tumor imaging agents.
Synthesis and biological evaluation of (18)F-labeled Fluoroethoxy tryptophan analogues as potential PET tumor imaging agents.
Molecular pharmaceutics (2014-07-06)
Aristeidis Chiotellis, Adrienne Muller, Linjing Mu, Claudia Keller, Roger Schibli, Stefanie D Krämer, Simon M Ametamey
ABSTRACT
As a continuation of our research efforts toward the development of tryptophan-based radiotracers for tumor imaging with positron emission tomography (PET), three new fluoroethoxy tryptophan analogues were synthesized and evaluated in vivo. These new tracers (namely, 4-(2-[(18)F]fluoroethoxy)-dl-tryptophan ([(18)F]4-FEHTP), 6-(2-[(18)F]fluoroethoxy)-dl-tryptophan ([(18)F]6-FEHTP), and 7-(2-[(18)F]fluoroethoxy)-dl-tryptophan ([(18)F]7-FEHTP) carry the fluoroethoxy side chain either at positions 4-, 6-, or 7- of the indole core. Reference compounds and precursors were synthesized by multistep approaches. Radiosynthesis was accomplished by no-carrier-added nucleophilic (18)F-fluorination following either an indirect approach (O-alkylation of the corresponding hydroxytryptophan with [(18)F]fluoroethyltosylate) or a direct approach (nucleophilic [(18)F] fluorination using a protected mesyl precursor). Radiochemical yields (decay corrected) for both methods were in the range of 10-18%. Small animal PET imaging with xenograft-bearing mice revealed the highest tumor/background ratio for [(18)F]6-FEHTP which, in a direct comparison, outperformed the other two tryptophan tracers and also the well-established tyrosine analogue O-(2-[(18)F]fluoroethyl)-l-tyrosine ([(18)F]l-FET). Investigation of the transport mechanism of [(18)F]6-FEHTP in small cell lung cancer cells (NCI-H69) revealed that it is most probably taken up exclusively via the large neutral amino acid transporter(s) (LAT).
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Tryptophan, European Pharmacopoeia (EP) Reference Standard