Merck
  • Identification of DEK as a potential therapeutic target for neuroendocrine prostate cancer.
Identification of DEK as a potential therapeutic target for neuroendocrine prostate cancer.
Oncotarget (2014-12-30)
Dong Lin, Xin Dong, Kendric Wang, Alexander W Wyatt, Francesco Crea, Hui Xue, Yuwei Wang, Rebecca Wu, Robert H Bell, Anne Haegert, Sonal Brahmbhatt, Antonio Hurtado-Coll, Peter W Gout, Ladan Fazli, Martin E Gleave, Colin C Collins, Yuzhuo Wang
ABSTRACT
Neuroendocrine prostate cancer (NEPC) is an aggressive subtype of prostate cancer which does not respond to hormone therapy. Research of NEPC has been hampered by a lack of clinically relevant in vivo models. Recently, we developed a first-in-field patient tissue-derived xenograft model of complete neuroendocrine transdifferentiation of prostate adenocarcinoma. By comparing gene expression profiles of a transplantable adenocarcinoma line (LTL331) and its NEPC subline (LTL331R), we identified DEK as a potential biomarker and therapeutic target for NEPC. In the present study, elevated DEK protein expression was observed in all NEPC xenograft models and clinical NEPC cases, as opposed to their benign counterparts (0%), hormonal naïve prostate cancer (2.45%) and castration-resistant prostate cancer (29.55%). Elevated DEK expression was found to be an independent clinical risk factor, associated with shorter disease-free survival of hormonal naïve prostate cancer patients. DEK silencing in PC-3 cells led to a marked reduction in cell proliferation, cell migration and invasion. The results suggest that DEK plays an important role in the progression of prostate cancer, especially to NEPC, and provides a potential biomarker to aid risk stratification of prostate cancer and a novel target for therapy of NEPC.
Sigma-Aldrich
MISSION® esiRNA, targeting human DEK
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dek
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)