Merck
  • Effects of total lignans from Eucommia ulmoides barks prevent bone loss in vivo and in vitro.
Effects of total lignans from Eucommia ulmoides barks prevent bone loss in vivo and in vitro.
Journal of ethnopharmacology (2014-05-03)
Rong Zhang, Ya-Lei Pan, Shi-Jie Hu, Xiang-He Kong, Wang Juan, Qi-Bing Mei
ABSTRACT
The present study systematically investigate the in vivo and in vitro effect of total lignans (TL) extracted from Eucommia ulmoides Oliv. barks on bone formation using ovariectomy rat model and primary cultures of rat osteoblasts. Eighty 3-month-old female Sprague-Dawley rats were used and randomly assigned into sham-operated group (SHAM) and five ovariectomy (OVX) subgroups, i.e. OVX with vehicle (OVX); OVX with 17Ī±-ethinylestradiol (E2, 25 Ī¼g/kg/day); OVX with TL of graded doses (20, 40, or 80 mg/kg/day). The treatment began 4 weeks after the surgery and lasted for 16 weeks. in vitro experiments were performed to determine the potential mechanisms of the anti-osteoporotic effect of TL. Treatment with TL significantly prevent OVX-induced decrease in biomechanical quality of femur such as maximum stress and Young׳s modulus. The mechanical changes were associated with the prevention of a further BMD decrease or even with some improvements in microarchitecture. TL inhibited BMD decrease in the femur caused by OVX, which was accompanied by a significant decrease in skeletal remodeling, as was evidenced by the decreased levels of the bone turnover markers. Ī¼CT analysis of the femoral metaphysis showed how to prevent the deterioration of trabecular microarchitecture. TL induced primary osteoblastic cell proliferation and differentiation, inhibition of osteoclastogenesis through an increase in osteoprotegrin (OPG) and a decrease in NF-ĪŗB ligand (RANKL) expression in vitro. We concluded that TL treatment can effectively suppress the loss of bone mass induced by OVX and in vitro evidence suggests this could be through actions on both osteoblasts and osteoclasts.
Sigma-Aldrich
Human CEACAM-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine