Merck
  • High EGFR gene copy number predicts poor outcome in triple-negative breast cancer.
High EGFR gene copy number predicts poor outcome in triple-negative breast cancer.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2014-01-11)
Heae Surng Park, Min Hye Jang, Eun Joo Kim, Hyun Jeong Kim, Hee Jin Lee, Yu Jung Kim, Jee Hyun Kim, Eunyoung Kang, Sung-Won Kim, In Ah Kim, So Yeon Park
ABSTRACT
Epidermal growth factor receptor (EGFR) is frequently overexpressed in triple-negative breast cancer and is emerging as a therapeutic target. EGFR gene copy number alteration and mutation are highly variable and scientists have been challenged to define their prognostic significance in triple-negative breast cancer. We examined EGFR protein expression, EGFR gene copy number alteration and mutation of exon 18 to 21 in 151 cases of triple-negative breast cancer and correlated these findings with clinical outcomes. In addition, intratumoral agreement of EGFR protein overexpression and gene copy number alteration was evaluated. EGFR overexpression was found in 97 of 151 cases (64%) and high EGFR gene copy number was detected in 50 cases (33%), including 3 gene amplification (2%) and 47 high polysomy (31%). Five EGFR mutations were detected in 4 of 151 cases (3%) and included G719A in exon 18 (n=1), V786M in exon 20 (n=1), and L858R in exon 21 (n=3). One case had two mutations (G719A and L858R). High EGFR copy number, but not EGFR mutation, correlated with EGFR protein overexpression. Intratumoral heterogeneity of EGFR protein overexpression and EGFR copy number alteration was not significant. In survival analyses, high EGFR copy number was found to be an independent prognostic factor for poor disease-free survival in patients with triple-negative breast cancer. Our findings showed that EGFR mutation was a rare event, but high EGFR copy number was relatively frequent and correlated with EGFR overexpression in triple-negative breast cancer. Moreover, high EGFR copy number was associated with poor clinical outcome in triple-negative breast cancer, suggesting that evaluation of EGFR copy number may be useful for predicting outcomes in patients with triple-negative breast cancer and for selecting patients for anti-EGFR-targeted therapy.
Product Description
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard