Merck
  • CXCL10-induced migration of adoptively transferred human natural killer cells toward solid tumors causes regression of tumor growth in vivo.
CXCL10-induced migration of adoptively transferred human natural killer cells toward solid tumors causes regression of tumor growth in vivo.
Cancer immunology, immunotherapy : CII (2014-10-27)
Erik Wennerberg, Veronika Kremer, Richard Childs, Andreas Lundqvist
ABSTRACT
Adoptive infusion of natural killer (NK) cells is being increasingly explored as a therapy in patients with cancer, although clinical responses are thus far limited to patients with hematological malignancies. Inadequate homing of infused NK cells to the tumor site represents a key factor that may explain the poor anti-tumor effect of NK cell therapy against solid tumors. One of the major players in the regulation of lymphocyte chemotaxis is the chemokine receptor chemokine (C-X-C motif) receptor 3 (CXCR3) which is expressed on activated NK cells and induces NK cell migration toward gradients of the chemokine (C-X-C motif) ligand (CXCL9, 10 and 11). Here, we show that ex vivo expansion of human NK cells results in a tenfold increased expression of the CXCR3 receptor compared with resting NK cells (p = 0.04). Consequently, these NK cells displayed an improved migratory capacity toward solid tumors, which was dependent on tumor-derived CXCL10. In xenograft models, adoptively transferred NK cells showed increased migration toward CXCL10-transfected melanoma tumors compared with CXCL10-negative wild-type tumors, resulting in significantly reduced tumor burden and increased survival (median survival 41 vs. 32 days, p = 0.03). Furthermore, administration of interferon-gamma locally in the tumor stimulated the production of CXCL10 in subcutaneous melanoma tumors resulting in increased infiltration of adoptively transferred CXCR3-positive expanded NK cells. Our findings demonstrate the importance of CXCL10-induced chemoattraction in the anti-tumor response of adoptively transferred expanded NK cells against solid melanoma tumors.
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)