Merck
Rapid selection of high-affinity binders using ribosome display.
Methods in molecular biology (Clifton, N.J.) (2011-11-19)
Birgit Dreier, Andreas Plückthun
ABSTRACT
Ribosome display has proven to be a powerful in vitro selection and evolution method for generating high-affinity binders from libraries of folded proteins. It has been successfully applied to single-chain Fv fragments of antibodies and alternative scaffolds, such as Designed Ankyrin Repeat Proteins (DARPins). High-affinity binders with new target specificity can be obtained from highly diverse DARPin libraries in only a few selection rounds. In this protocol, the selection from the library and the process of affinity maturation and off-rate selection are explained in detail.
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)