Merck
  • miR-490-3p modulates cell growth and epithelial to mesenchymal transition of hepatocellular carcinoma cells by targeting endoplasmic reticulum-Golgi intermediate compartment protein 3 (ERGIC3).
miR-490-3p modulates cell growth and epithelial to mesenchymal transition of hepatocellular carcinoma cells by targeting endoplasmic reticulum-Golgi intermediate compartment protein 3 (ERGIC3).
The Journal of biological chemistry (2012-12-06)
Ling-yun Zhang, Min Liu, Xin Li, Hua Tang
ABSTRACT
MicroRNAs (miRNAs) are considered to be regulators of various biological processes in cancers, including the epithelial to mesenchymal transition (EMT), which is a key factor in cancer metastasis. In this study, we aimed to clarify the potential roles of miR-490-3p in hepatocellular carcinoma (HCC) cells. Using real-time quantitative RT-PCR, we discovered that miR-490-3p was up-regulated in HCC tissues and cells compared with the adjacent non-tumor tissues and normal cells. We also found that overexpression of miR-490-3p led to an increase in cell proliferation, migration, and invasion abilities and that it contributed to EMT. The inhibition of miR-490-3p had the opposite effect on the cells. We identified ERGIC3 (endoplasmic reticulum-Golgi intermediate compartment protein 3) as a direct target gene for miR-490-3p. Unlike most miRNA-mRNA interactions, miR-490-3p increased ERGIC3 mRNA and protein levels as well as the intensity of expression of the EGFP reporter gene controlled by the 3'-UTR of ERGIC3 mRNA. The up-regulation by miR-490-3p also required the participation of Ago2. The inhibition of miR-490-3p reduced the expression of ERGIC3. Overexpression of ERGIC3 led to the same effect on HCC cells as miR-490-3p overexpression, including EMT. Importantly, silencing ERGIC3 reversed the cellular responses mediated by miR-490-3p overexpression. In conclusion, our study indicated for the first time that miR-490-3p functioned like an oncogenic miRNA in HCC cells and that the inhibition of miR-490-3p might provide an potential treatment approach for HCC patients.