Merck
  • Antagonistic actions of Rcor proteins regulate LSD1 activity and cellular differentiation.
Antagonistic actions of Rcor proteins regulate LSD1 activity and cellular differentiation.
Proceedings of the National Academy of Sciences of the United States of America (2014-05-21)
Ghanshyam Upadhyay, Asif H Chowdhury, Bharat Vaidyanathan, David Kim, Shireen Saleque
ABSTRACT
Lysine-specific demethylase 1 (LSD1) demethylates nucleosomal histone H3 lysine 4 (H3K4) residues in collaboration with the corepressor CoREST/REST corepressor 1 (Rcor1) and regulates cell fates by epigenetically repressing gene targets. The balanced regulation of this demethylase, if any, is however unknown. We now demonstrate the actions of two other Rcor paralogs, Rcor2 and Rcor3, in regulating LSD1 enzymatic activity and biological function in hematopoietic cells. All three Rcor proteins interact with LSD1 and with the erythro-megakaryocytic transcription factor growth factor independence (Gfi)1b; however, whereas Rcor2, like Rcor1, facilitates LSD1-mediated nucleosomal demethylation, Rcor3 competitively inhibits this process. Appending the SANT2 domain of Rcor1 to Rcor3 confers the ability to facilitate LSD1-mediated demethylation on the chimeric Rcor protein. Consistent with their biochemical activities, endogenous Rcor1, Rcor2, and LSD1 promote differentiation, whereas Rcor3 opposes these processes. Recruitment of Rcor3 to cognate gene targets by Gfi1b and LSD1 leads to inhibition of H3K4 demethylation of chromatin and transcriptional derepression of these loci. Remarkably, profound alterations in Rcor1/3 levels during erythroid versus megakaryocytic differentiation potentiate antagonistic outcomes. In mature erythroid cells, a strong upsurge in Rcor3 and a sharp decline in Rcor1 levels counteract LSD1/Rcor1/2-mediated differentiation. In contrast, the opposite changes in Rcor1/3 levels in megakaryocytes favor differentiation and likely maintain homeostasis between these lineages. Overall, our results identify Rcor3 as a natural inhibitor of LSD1 and highlight a dual mechanism of regulating the enzymatic activity and restraining the epigenetic impact of this robust demethylase during hematopoietic differentiation.
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Aof2
Sigma-Aldrich
Anti-Histone H3 Antibody, clone 6.6.2, clone 6.6.2, Upstate®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human KDM1A