Merck
  • Asymmetric synthesis of primary amines via the spiroborate-catalyzed borane reduction of oxime ethers.
Asymmetric synthesis of primary amines via the spiroborate-catalyzed borane reduction of oxime ethers.
Organic letters (2007-04-03)
Xiaogen Huang, Margarita Ortiz-Marciales, Kun Huang, Viatcheslav Stepanenko, Francisco G Merced, Angel M Ayala, Wildeliz Correa, Melvin De Jesús
ABSTRACT
[reaction: see text] The enantioselective borane reduction of O-benzyloxime ethers to primary amines was studied under catalytic conditions using the spiroborate esters 5-10 derived from nonracemic 1,2-amino alcohols and ethylene glycol. Effective catalytic conditions were achieved using only 10% of catalyst 5 derived from diphenylvalinol in dioxane at 0 degrees C resulting in complete conversion to the corresponding primary amine in up to 99% ee.
Sigma-Aldrich
N-Isopropyl-N-methyl-tert-butylamine, 98%