Merck
  • Continuous culture of Methanococcus maripaludis under defined nutrient conditions.
Continuous culture of Methanococcus maripaludis under defined nutrient conditions.
FEMS microbiology letters (2004-09-01)
Andrew K Haydock, Iris Porat, William B Whitman, John A Leigh
ABSTRACT
To study global regulation in the methanogenic archaeon Methanococcus maripaludis, we devised a system for steady-state growth in chemostats. New Brunswick Bioflo 110 bioreactors were equipped with controlled delivery of hydrogen, nitrogen, carbon dioxide, hydrogen sulfide, and anaerobic medium. We determined conditions and media compositions for growth with three different limiting nutrients, hydrogen, phosphate, and leucine. To investigate leucine limitation we constructed and characterized a mutant in the leuA gene for 2-isopropylmalate synthase, demonstrating for the first time the function of this gene in the Archaea. Steady state specific growth rates in these studies ranged from 0.042 to 0.24 h(-1). Plots of culture density vs. growth rate for each condition showed the behavior predicted by growth modeling. The results show that growth behavior is normal and reproducible and validate the use of the chemostat system for metabolic and global regulation studies in M. maripaludis.