Merck
  • Structure-in vitro activity relationships of pentamidine analogues and dication-substituted bis-benzimidazoles as new antifungal agents.
Structure-in vitro activity relationships of pentamidine analogues and dication-substituted bis-benzimidazoles as new antifungal agents.
Antimicrobial agents and chemotherapy (1998-10-03)
M Del Poeta, W A Schell, C C Dykstra, S Jones, R R Tidwell, A Czarny, M Bajic, A Kumar, D Boykin, J R Perfect
ABSTRACT
Twenty analogues of pentamidine, 7 primary metabolites of pentamidine, and 30 dicationic substituted bis-benzimidazoles were screened for their inhibitory and fungicidal activities against Candida albicans and Cryptococcus neoformans. A majority of the compounds had MICs at which 80% of the strains were inhibited (MIC80s) comparable to those of amphotericin B and fluconazole. Unlike fluconazole, many of these compounds were found to have potent fungicidal activity. The most potent compound against C. albicans had an MIC80 of </=0.09 microg/ml, and the most potent compound against C. neoformans had an MIC80 of 0.19 microg/ml. Selected compounds were also found to be active against Aspergillus fumigatus, Fusarium solani, Candida species other than C. albicans, and fluconazole-resistant strains of C. albicans and C. neoformans. It is clear from the data presented here that further studies on the structure-activity relationships, mechanisms of action and toxicities, and in vivo efficacies of these compounds are warranted to determine their clinical potential.
Sigma-Aldrich
2,5-Thiophenedicarboxaldehyde, 99%