Merck
What are the current clinical issues in wear and tribocorrosion?
Clinical orthopaedics and related research (2014-04-09)
Daniel J Berry, Matthew P Abdel, John J Callaghan
ABSTRACT
Wear and corrosion in joint arthroplasty are important causes of failure. From the standpoint of current clinical importance, there are four main categories of wear and tribocorrosion: polyethylene wear, ceramic-on-ceramic (CoC) bearing wear, metal-on-metal (MoM) bearing wear, and taper tribocorrosion. Recently, problems with wear in the knee have become less prominent as have many issues with hip polyethylene (PE) bearings resulting from the success of crosslinked PE. However, MoM articulations and taper tribocorrosion have been associated with soft tissue inflammatory responses, and as a result, they have become prominent clinical concerns. WHERE ARE WE NOW?: For PE wear in the hip, several advances include improved locking mechanisms and data supporting highly crosslinked polyethylenes (HXLPE). Edge-loading in CoC articulations can contribute to stripe wear and subsequent squeaking. For MoM articulations, the relationship of wear-to-edge loading, sensitivity to component positioning, typical soft tissue response, and use of imaging is increasingly understood. Taper tribocorrosion (from femoral head-neck junctions and other modular elements) and associated soft tissue inflammatory responses appear to be serious clinical issues that are not fully understood. WHERE DO WE NEED TO GO?: In the knee, clinical concerns remain with the efficacy of HXLPE, modular connections, and metal allergies. For PE wear in the hip, concerns remain regarding how to increase crosslinking of PE while minimizing PE fractures. With CoC articulations, questions remain on how to prevent noises, chipping, and impingement and if enhanced designs can contribute to improved results. For MoM articulations, we need to improve imaging tests for soft tissue reactions, determine best practices in terms of monitoring protocols, and better define if, how, and when to act on serum metal levels. For taper tribocorrosion, we need to use modularity wisely and also understand how to improve tapers and materials in the future. For patients at risk for tribocorrosion, we need to define realistic diagnostic and monitoring protocols. We also need to enhance revision methods, and the threshold of acceptable soft tissue damage, to minimize complications associated with soft tissue damage such as hip instability. HOW DO WE GET THERE?: HXLPE and other bearing surfaces will likely continue to be refined. We need to develop tapers with more resistance to tribocorrosion through improved understanding of the manufacturing process and ongoing engineering improvements. Revision procedures for wear and tribocorrosion can be enhanced by determining when partial component retention is appropriate and how best to manage soft tissue damage. For CoC articulations, enhanced designs are required to minimize noises, chipping, and impingement. Importantly, we must continue to promote and analyze joint replacement registries to identify early failures and analyze long-term successes.
Product Description
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC