Merck
  • Aminopeptidase-N/CD13 is a potential proapoptotic target in human myeloid tumor cells.
Aminopeptidase-N/CD13 is a potential proapoptotic target in human myeloid tumor cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2011-05-14)
Marion Piedfer, Daniel Dauzonne, Ruoping Tang, Juliette N'Guyen, Christian Billard, Brigitte Bauvois
ABSTRACT
The transmembrane metalloprotease aminopeptidase-N (APN)/CD13 is overexpressed in various solid and hematological malignancies in humans, including acute myeloid leukemia (AML) and is thought to influence tumor progression. Here, we investigated the contribution of APN/CD13 to the regulation of growth and survival processes in AML cells in vitro. Anti-CD13 monoclonal antibodies MY7 and SJ1D1 (which do not inhibit APN activity) and WM15 (an APN-blocking antibody) inhibited the growth of the AML cell line U937 and induced apoptosis, as evidenced by cell accumulation in the sub-G(1) phase, DNA fragmentation, and phosphatidylserine externalization. Isotype-matched IgG1 and the APN/CD13 enzymatic inhibitors bestatin and 2',3-dinitroflavone-8-acetic acid, were ineffective. Internalization of CD13-MY7 complex into cells was followed by mitochondrial membrane depolarization, Bcl-2 and Mcl-1 down-regulation, Bax up-regulation, caspase-9, caspase-8, and caspase-3 activation, and cleavage of the caspase substrate PARP-1. The broad-spectrum caspase inhibitor Z-VAD-fmk and the caspase-9- and caspase-8-specific inhibitors significantly attenuated apoptosis. CD13 ligation also induced apoptosis and PARP-1 cleavage in primary AML blasts, whereas normal blood cells were not affected. Overall, these data provide new evidence that CD13 can serve as a target for inducing caspase-dependent apoptosis in AML (independently of its APN activity). These findings may have implications for tumor biology and treatment.
Sigma-Aldrich
Bestatin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
CD13 (SP187) Rabbit Monoclonal Antibody