- Expression of dishevelled gene in Hirschsprung's disease.
Expression of dishevelled gene in Hirschsprung's disease.
Hirschsprung's disease (HSCR) is a congenital disorder of the enteric nervous system and is characterized by an absence of enteric ganglion cells in terminal regions of the gut during development. Dishevelled (DVL) protein is a cytoplasmic protein which plays pivotal roles in the embryonic development. In this study, we explore the cause of HSCR by studying the expression of DVL-1 and DVL-3 genes and their proteins in the aganglionic segment and the ganglionic segment of colon in HSCR patients. Specimen of aganglionic segment and ganglionic segment of colon in 50 cases of HSCR patients. Expression levels of mRNA and proteins of DVL-1 and DVL-3 were confirmed by quantitative real-time PCR (qRT-PCR), western blot and immunohistochemistry staining between the aganglionic segment and the ganglionic segment of colon in HSCR patients. The mRNA expression of DVL-1 and DVL-3 were 2.06 fold and 3.12 fold in the aganglionic segment colon tissues compared to the ganglionic segment, respectively. Similarly, the proteins expression of DVL-1 and DVL-3 were higher (39.71 ± 4.53 vs and 53.90 ± 6.79 vs) in the aganglionic segment colon tissues than in the ganglionic segment (15.01 ± 2.66 and 20.13 ± 3.63) by western blot. Besides, immunohistochemical staining showed that DVL-1 and DVL-3 have a significant increase in mucous and submucous layers from aganglionic colon segments compared with ganglionic segments. The study showed an association of DVL-1 and DVL-3 with HSCR, it may play an important role in the pathogenesis of HSCR.