Merck
  • Pseudomonas hussainii sp. nov., isolated from droppings of a seashore bird, and emended descriptions of Pseudomonas pohangensis, Pseudomonas benzenivorans and Pseudomonas segetis.
Pseudomonas hussainii sp. nov., isolated from droppings of a seashore bird, and emended descriptions of Pseudomonas pohangensis, Pseudomonas benzenivorans and Pseudomonas segetis.
International journal of systematic and evolutionary microbiology (2014-04-20)
Asif Hameed, Mariyam Shahina, Shih-Yao Lin, You-Cheng Liu, Chiu-Chung Young
ABSTRACT
Two Gram-staining-negative, aerobic, rod-shaped, non-spore-forming bacterial strains that are motile by a monopolar flagellum, designated CC-AMH-11(T) and CC-AMHZ-5, were isolated from droppings of a seashore bird off the coast of Hualien, Taiwan. The strains showed 99.7% mutual pairwise 16S rRNA gene sequence similarity, while exhibiting <96.2% sequence similarity to strains of other species of the genus Pseudomonas (95.7-95.9% similarity with type species, Pseudomonas aeruginosa LMG 1242T), and formed a distinct co-phyletic lineage in the phylogenetic trees. The common major fatty acids (>5% of the total) were C18 : 1ω7c and/or C18 : 1ω6c (summed feature 8), C16 : 1ω6c and/or C16 : 1ω7c (summed feature 3), C16 : 0 and C12 : 0. Phosphatidylethanolamine, phosphatidylglycerol, diphosphatidylglycerol, phosphatidylserine, an unidentified lipid and an unidentified phospholipid were detected as common polar lipids. The DNA G+C contents of strains CC-AMH-11(T) and CC-AMHZ-5 were 61.1 and 61.6 mol%, respectively. The common major respiratory quinone was ubiquinone 9 (Q-9), and the predominant polyamine was putrescine. The DNA-DNA hybridization obtained between the two strains was 79.0% (reciprocal value 89.4% using CC-AMHZ-5 DNA as the probe). The very high 16S rRNA gene sequence similarity and DNA-DNA relatedness and the poorly distinguishable phenotypic features witnessed between CC-AMH-11(T) and CC-AMHZ-5 suggested unambiguously that they are two distinct strains of a single genomic species. However, the strains also showed several genotypic and phenotypic characteristics that distinguished them from other closely related species of Pseudomonas. Thus, the strains are proposed to represent a novel species of Pseudomonas, for which the name Pseudomonas hussainii sp. nov. is proposed. The type strain is CC-AMH-11(T) ( = JCM 19513(T) = BCRC 80696(T)); a second strain of the same species is CC-AMHZ-5 ( = JCM 19512 = BCRC 80697). In addition, emended descriptions of the species Pseudomonas pohangensis, Pseudomonas benzenivorans and Pseudomonas segetis are also proposed.
Product Description
USP
Ubidecarenone Related Compound A, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Coenzyme Q9, ≥96.0% (HPLC)
Ubidecarenone impurity D, European Pharmacopoeia (EP) Reference Standard