Merck
  • Azo group-assisted nucleophilic aromatic substitutions in haloarene derivatives: preparation of substituted 1-iodo-2,6-bispropylthiobenzenes.
Azo group-assisted nucleophilic aromatic substitutions in haloarene derivatives: preparation of substituted 1-iodo-2,6-bispropylthiobenzenes.
The Journal of organic chemistry (2004-04-03)
Jason T Manka, Virginia C McKenzie, Piotr Kaszynski
ABSTRACT
Aryldiazo substituents were used in nucleophilic aromatic substitution reactions of halogens. The Ph-N=N- group activates ortho fluorine atoms toward alkylthiolation under mild conditions. In contrast, the Me(2)N-C(6)H(4)-N=N- group has virtually no activation effect in nucleophilic aromatic substitution, and serves as a "neutral" mask for the amino group. The Ph-N=N- group was efficiently introduced by diazo coupling of aryllithium with dry PhN(2)(+)BF(4)(-) salt.
Sigma-Aldrich
1-Chloro-2,4-difluorobenzene, 98%