Merck
  • Regioselective functionalization of the thiazole scaffold using TMPMgCl·LiCl and TMP2Zn·2MgCl2·2LiCl.
Regioselective functionalization of the thiazole scaffold using TMPMgCl·LiCl and TMP2Zn·2MgCl2·2LiCl.
The Journal of organic chemistry (2011-07-09)
Cora Dunst, Paul Knochel
ABSTRACT
A general method for the synthesis of 2,4,5-trisubstituted thiazoles has been developed. Starting from commercially available 2-bromothiazole, successive metalations using TMPMgCl·LiCl or TMP(2)Zn·2MgCl(2)·2LiCl lead to the corresponding magnesated or zincated thiazoles which readily react with various electrophiles providing highly functionalized thiazoles.
Sigma-Aldrich
2-Bromothiazole, 98%