Merck
  • Syntheses and spectroscopic studies of spirobifluorene-bridged bipolar systems; photoinduced electron transfer reactions.
Syntheses and spectroscopic studies of spirobifluorene-bridged bipolar systems; photoinduced electron transfer reactions.
Chemical communications (Cambridge, England) (2002-12-14)
Yuh-Yih Chien, Ken-Tsung Wong, Pi-Tai Chou, Yi-Ming Cheng
ABSTRACT
Some 9,9'-spirobifluorene-bridged bipolar systems 1-3 containing 1,3,4-oxadiazole-conjugated oligoaryl and triarylamine moieties have been synthesized, in which 1 exhibits remarkable solvent-polarity dependent fluorescence properties due to a highly efficient photoinduced electron transfer reaction.
Sigma-Aldrich
4-tert-Butylbenzoyl chloride, 98%