Merck
Metabolically Stable tert-Butyl Replacement.
ACS medicinal chemistry letters (2014-06-06)
David Barnes-Seeman, Monish Jain, Leslie Bell, Suzie Ferreira, Scott Cohen, Xiao-Hui Chen, Jakal Amin, Brad Snodgrass, Panos Hatsis
ABSTRACT
Susceptibility to metabolism is a common issue with the tert-butyl group on compounds of medicinal interest. We demonstrate an approach of removing all the fully sp(3) C-Hs from a tert-butyl group: replacing some C-Hs with C-Fs and increasing the s-character of the remaining C-Hs. This approach gave a trifluoromethylcyclopropyl group, which increased metabolic stability. Trifluoromethylcyclopropyl-containing analogues had consistently higher metabolic stability in vitro and in vivo compared to their tert-butyl-containing counterparts.
Sigma-Aldrich
Sodium 1-(trifluoromethyl)cyclopropanesulfinate