MilliporeSigma
  • Antitumor agents. 217. Curcumin analogues as novel androgen receptor antagonists with potential as anti-prostate cancer agents.
Antitumor agents. 217. Curcumin analogues as novel androgen receptor antagonists with potential as anti-prostate cancer agents.
Journal of medicinal chemistry (2002-11-01)
Hironori Ohtsu, Zhiyan Xiao, Junko Ishida, Masahiro Nagai, Hui-Kang Wang, Hideji Itokawa, Ching-Yuan Su, Charles Shih, Tzuying Chiang, Eugene Chang, YiFen Lee, Meng-Yin Tsai, Chawnshang Chang, Kuo-Hsiung Lee
ABSTRACT
A number of curcumin analogues were prepared and evaluated as potential androgen receptor antagonists against two human prostate cancer cell lines, PC-3 and DU-145, in the presence of androgen receptor (AR) and androgen receptor coactivator, ARA70. Compounds 4 [5-hydroxy-1,7-bis(3,4-dimethoxyphenyl)-1,4,6-heptatrien-3-one], 20 [5-hydroxy-1,7-bis[3-methoxy-4-(methoxycarbonylmethoxy)phenyl]-1,4,6-heptatrien-3-one], 22 [7-(4-hydroxy-3-methoxyphenyl)-4-[3-(4-hydroxy-3-methoxyphenyl)acryloyl]-5-oxohepta-4,6-dienoic acid ethyl ester], 23 [7-(4-hydroxy-3-methoxyphenyl)-4-[3-(4-hydroxy-3-methoxyphenyl)acryloyl]5-oxohepta-4,6-dienoic acid], and 39 [bis(3,4-dimethoxyphenyl)-1,3-propanedione] showed potent antiandrogenic activities and were superior to hydroxyflutamide, which is the currently available antiandrogen for the treatment of prostate cancer. Structure-activity relationship (SAR) studies indicated that the bis(3,4-dimethoxyphenyl) moieties, the conjugated beta-diketone moiety, and the intramolecular symmetry of the molecules seem to be important factors related to antiandrogenic activity. The data further suggest that the coplanarity of the beta-diketone moiety and the presence of a strong hydrogen bond donor group were also crucial for the antiandrogenic activity, which is consistent with previous SAR results for hydroxyflutamide analogues. When the pharmacophoric elements of dihydrotestosterone (DHT) and compound 4 are superposed, the resulting construct implies that the curcumin analogues may function as a 17alpha-substituted DHT. Compounds 4, 20, 22, 23, and 39 have been identified as a new class of antiandrogen agents, and these compounds or their new synthetic analogues could be developed into clinical trial candidates to control androgen receptor-mediated prostate cancer growth.