Merck
Melamine contamination.
Analytical and bioanalytical chemistry (2009-08-12)
Yu-Chang Tyan, Ming-Hui Yang, Shiang-Bin Jong, Chih-Kuang Wang, Jentaie Shiea
ABSTRACT
In the summer of 2008, serious illnesses and deaths of babies in China were linked to melamine-tainted powdered infant formula. Melamine contains several metabolites, such as ammeline, ammelide, and cyanuric acid, and has been used for the adulteration of foods or milk to increase their apparent protein content. It is assumed that melamine and its metabolites are absorbed in the gastrointestinal tract, and precipitate in the kidney to form crystals. A new tolerable daily intake of 0.2 mg kg(-1) body weight was adapted by the World Health Organization in 2008. This paper reviews the variety of analytical methods that have been used for the analysis of melamine in food. The limit of detection of these various methods is 0.05-100 ppm. The maximum acceptable concentration in food has been set at 50 ppb by the US FDA. A fast and ultrasensitive procedure for screening, detection, and characterization of melamine and its derivative compounds needs to be established. Currently, mass-spectrometry technologies provide an alternative to derivatization for regulatory analysis of food.
Product Description
Supelco
Melamine, analytical standard
USP
Melamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Melamine, 99%
Supelco
Melamine, Pharmaceutical Secondary Standard; Certified Reference Material