Merck
  • The cell cycle as a target for cancer therapy: basic and clinical findings with the small molecule inhibitors flavopiridol and UCN-01.
The cell cycle as a target for cancer therapy: basic and clinical findings with the small molecule inhibitors flavopiridol and UCN-01.
The oncologist (2002-08-08)
Adrian M Senderowicz
ABSTRACT
Many tumor types are associated with genetic changes in the retinoblastoma pathway, leading to hyperactivation of cyclin-dependent kinases and incorrect progression through the cell cycle. Small-molecule cyclin-dependent kinase inhibitors are being developed as therapeutic agents. Of these, flavopiridol and UCN-01 are being explored in cancer patients in phase I and phase II clinical trials, both as single agents and in combination with conventional chemotherapeutic agents. The present article discusses the mechanisms of action of flavopiridol and UCN-01 as well as the outcome of clinical trials with these novel agents.
Sigma-Aldrich
UCN-01, ≥97% (HPLC), powder