Merck
  • Hepatitis C virus triggers mitochondrial fission and attenuates apoptosis to promote viral persistence.
Hepatitis C virus triggers mitochondrial fission and attenuates apoptosis to promote viral persistence.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-16)
Seong-Jun Kim, Gulam H Syed, Mohsin Khan, Wei-Wei Chiu, Muhammad A Sohail, Robert G Gish, Aleem Siddiqui
ABSTRACT
Mitochondrial dynamics is crucial for the regulation of cell homeostasis. Our recent findings suggest that hepatitis C virus (HCV) promotes Parkin-mediated elimination of damaged mitochondria (mitophagy). Here we show that HCV perturbs mitochondrial dynamics by promoting mitochondrial fission followed by mitophagy, which attenuates HCV-induced apoptosis. HCV infection stimulated expression of dynamin-related protein 1 (Drp1) and its mitochondrial receptor, mitochondrial fission factor. HCV further induced the phosphorylation of Drp1 (Ser616) and caused its subsequent translocation to the mitochondria, followed by mitophagy. Interference of HCV-induced mitochondrial fission and mitophagy by Drp1 silencing suppressed HCV secretion, with a concomitant decrease in cellular glycolysis and ATP levels, as well as enhanced innate immune signaling. More importantly, silencing Drp1 or Parkin caused significant increase in apoptotic signaling, evidenced by increased cytochrome C release from mitochondria, caspase 3 activity, and cleavage of poly(ADP-ribose) polymerase. These results suggest that HCV-induced mitochondrial fission and mitophagy serve to attenuate apoptosis and may contribute to persistent HCV infection.
Sigma-Aldrich
O-Phospho-DL-serine, ≥98.0% (NT)