Merck
  • Health effects of ambient air pollution: do different methods for estimating exposure lead to different results?
Health effects of ambient air pollution: do different methods for estimating exposure lead to different results?
Environment international (2014-03-07)
Yann Sellier, Julien Galineau, Agnes Hulin, Fabrice Caini, Nathalie Marquis, Vladislav Navel, Sebastien Bottagisi, Lise Giorgis-Allemand, Claire Jacquier, Remy Slama, Johanna Lepeule
ABSTRACT
Spatially resolved exposure models are increasingly used in epidemiology. We previously reported that, although exhibiting a moderate correlation, pregnancy nitrogen dioxide (NO2) levels estimated by the nearest air quality monitoring station (AQMS) model and a geostatistical model, showed similar associations with infant birth weight. We extended this study by comparing a total of four exposure models, including two highly spatially resolved models: a land-use regression (LUR) model and a dispersion model. Comparisons were made in terms of predicted NO2 and particle (aerodynamic diameter<10 μm, PM10) exposure and adjusted association with birth weight. The four exposure models were implemented in two French metropolitan areas where 1026 pregnant women were followed as part of the EDEN mother-child cohort. Correlations between model predictions were high (≥ 0.70), except for NO2 between the AQMS and both the LUR (r = 0.54) and dispersion models (r = 0.63). Spatial variations as estimated by the AQMS model were greater for NO2 (95%) than for PM10 (22%). The direction of effect estimates of NO2 on birth weight varied according to the exposure model, while PM10 effect estimates were more consistent across exposure models. For PM10, highly spatially resolved exposure model agreed with the poor spatial resolution AQMS model in terms of estimated pollutant levels and health effects. For more spatially heterogeneous pollutants like NO2, although predicted levels from spatially resolved models (all but AQMS) agreed with each other, our results suggest that some may disagree with each other as well as with the AQMS regarding the direction of the estimated health effects.
Sigma-Aldrich
Nitrogen dioxide, ≥99.5%