Merck
  • Oxypurinol directly and immediately activates the drug-specific T cells via the preferential use of HLA-B*58:01.
Oxypurinol directly and immediately activates the drug-specific T cells via the preferential use of HLA-B*58:01.
Journal of immunology (Baltimore, Md. : 1950) (2014-03-05)
James Yun, Maria J Marcaida, Klara K Eriksson, Heidi Jamin, Stefano Fontana, Werner J Pichler, Daniel Yerly
ABSTRACT
Allopurinol (ALP) hypersensitivity is a major cause of severe cutaneous adverse reactions and is strongly associated with the HLA-B*58:01 allele. However, it can occur in the absence of this allele with identical clinical manifestations. The immune mechanism of ALP-induced severe cutaneous adverse reactions is poorly understood, and the T cell-reactivity pattern in patients with or without the HLA-B*58:01 allele is not known. To understand the interactions among the drug, HLA, and TCR, we generated T cell lines that react to ALP or its metabolite oxypurinol (OXP) from HLA-B*58:01(+) and HLA-B*58:01(-) donors and assessed their reactivity. ALP/OXP-specific T cells reacted immediately to the addition of the drugs and bypassed intracellular Ag processing, which is consistent with the "pharmacological interaction with immune receptors" (p-i) concept. This direct activation occurred regardless of HLA-B*58:01 status. Although most OXP-specific T cells from HLA-B*58:01(+) donors were restricted by the HLA-B*58:01 molecule for drug recognition, ALP-specific T cells also were restricted to other MHC class I molecules. This can be explained by in silico docking data that suggest that OXP binds to the peptide-binding groove of HLA-B*58:01 with higher affinity. The ensuing T cell responses elicited by ALP or OXP were not limited to particular TCR Vβ repertoires. We conclude that the drug-specific T cells are activated by OXP bound to HLA-B*58:01 through the p-i mechanism.
Product Description
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxypurinol, ≥98% (HPLC)
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor