Merck
  • Human FEM1B is required for Rad9 recruitment and CHK1 activation in response to replication stress.
Human FEM1B is required for Rad9 recruitment and CHK1 activation in response to replication stress.
Oncogene (2009-03-31)
T-P Sun, S-Y Shieh
ABSTRACT
Human checkpoint kinase 1 (CHK1) is an essential kinase required to preserve genome stability, and is activated by DNA replication blockage through the ataxia-telangiectasia-mutated-and-Rad3-related (ATR)/ATRIP-signaling pathway. In this report, we show that a novel CHK1-interacting protein, FEM1B (human homologue of the Caenorhabditis elegans sex determination fem1 protein), identified by a yeast two-hybrid screen, is involved in the activation of CHK1 by replication stress. Depletion of FEM1B by small interfering RNA in cancer cells impairs the activation of CHK1 kinase activity and attenuates the induction of CHK1 Ser345 phosphorylation upon replication interference. It is to be noted that, CHK2 Thr68 phosphorylation is not altered by FEM1B downregulation. By fractionation, we further demonstrated that FEM1B is able to associate with chromatin, and such association facilitates chromatin loading of the Rad9 protein. Consistently, ATR activity is poorly maintained in FEM1B knockdown cells; and FEM1B-ablated cells are as sensitive to replication block as CHK1-depleted cells. Our study has uncovered an adaptor protein FEM1B, which acts as a bridge linking CHK1 and Rad9, thus facilitating checkpoint signaling induced by replication stress.
Sigma-Aldrich
Anti-FEM1B (AB1) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution