Merck
  • 2-Methoxybenzoyl phosphate: a new substrate for continuous fluorimetric and spectrophotometric acyl phosphatase assays.
2-Methoxybenzoyl phosphate: a new substrate for continuous fluorimetric and spectrophotometric acyl phosphatase assays.
Experientia (1995-01-15)
P Paoli, G Camici, G Manao, G Ramponi
ABSTRACT
A new aromatic acyl phosphate, 2-methoxybenzoyl phosphate, has been synthesized. The compound shows an intrinsic fluorescence; it displays an intense emission band at 390 nm upon excitation in the near UV region. This band practically disappears after hydrolysis of the product. On the other hand, the product displays differences in the near UV absorption spectra measured before and after hydrolysis. The delta epsilon at 301 nm is 2720 M-1 cm-1, a value that is 4.3-fold higher than that of benzoyl phosphate (the usual substrate for acylphosphatase assay) at 283 nm. The main kinetic parameters of three different acylphosphatase molecular forms (the muscular isoenzyme and two subtypes of the organ common isoenzyme) were determined using both benzoyl phosphate and 2-methoxybenzoyl phosphate as substrates, and then compared. These kinetic data and the UV absorption and fluorescence properties of 2-methoxybenzoyl phosphate suggest that this compound has better substrate features than benzoyl phosphate, and can be used for both high sensitivity continuous fluorimetric and UV absorption spectrophotometric assays of acylphosphatase.
Sigma-Aldrich
2-Methoxybenzoyl chloride, 97%