- Novel liquid chromatography tandem mass spectrometry (LC-MS/MS) methods for measuring steroids.
Novel liquid chromatography tandem mass spectrometry (LC-MS/MS) methods for measuring steroids.
Liquid chromatography tandem mass spectrometry (LC-MS/MS) is increasingly becoming the method of choice for steroid hormone measurements due to small sample volumes, fast analysis times and improved specificity compared to immunoassays. Achievement of demanding analytical targets for steroid analysis is now becoming possible because of improvements in sample preparation technology, liquid chromatography column technology and mass spectrometer design. The most popular sample treatment strategies comprise protein precipitation (PP), solid-phase extraction (SLE) and liquid-liquid extraction (LLE). Modern liquid chromatography columns can ensure the adequate separation of isobaric compounds e.g. 21 Deoxycortisol, 11 Deoxycortisol and Corticosterone. The most appropriate method may be chosen to improve assay sensitivity by reducing matrix effects (LLE, SPE) or simplicity and speed (PP). Specific examples of some clinically important steroids including oestradiol, aldosterone, renin, serum cortisol, salivary cortisol and salivary testosterone will be described.