Merck
  • In vitro screening of carcinogenesis inhibitors acting by inhibition of microsomal polycyclic aromatic hydrocarbon activation.
In vitro screening of carcinogenesis inhibitors acting by inhibition of microsomal polycyclic aromatic hydrocarbon activation.
Neoplasma (1987-01-01)
C Safirman, G Stoica, F Lupu, N Voiculetz, I Niculescu-Duvaz
ABSTRACT
A procedure for in vitro prescreening of carcinogenesis inhibitors acting by the inhibition of the microsomal activation systems is described. It consists of measuring the protection conferred by the investigated chemicals against DNA binding of ultimate carcinogens resulted during G(3H)B[a]P activation. This rapid and reproducible procedure is able to select only carcinogenesis inhibitors which interact with the cytochrome P-450 dependent microsomal enzymes being specific in this respect. The effectiveness of 23 compounds (mainly antioxidants) was tested and the results compared with literary data concerning their activity. The concordance was satisfactory.
Product Description
USP
2-tert-Butyl-4-hydroxyanisole, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.