Merck
  • Effects of protein binding on a lipid bilayer containing local anesthetic articaine, and the potential of mean force calculation: a molecular dynamics simulation approach.
Effects of protein binding on a lipid bilayer containing local anesthetic articaine, and the potential of mean force calculation: a molecular dynamics simulation approach.
Journal of molecular modeling (2013-06-27)
Sepideh Amjad-Iranagh, Abbas Yousefpour, Parto Haghighi, Hamid Modarress
ABSTRACT
Articaine, as a local anesthetic drug has been simulated in neutral and charged forms, and its interaction with the dimyristoylphosphatidylcholine (DMPC) lipid bilayer membrane is investigated by molecular dynamics simulation using GROMACS software. In order to obtain the optimum location of the drug molecules, as they penetrate into the membrane, umbrella sampling is applied and the free energy is calculated. The effect of protein binding to DMPC membrane on the process of drug diffusion through the membrane is considered. Five simulation systems are designed and by applying the potential of mean force, the molecular dynamics simulation on the system is performed. In light of the obtained results, the electrostatic potential, variation of lipid bilayer's order parameter and the diffusion coefficient of drug are discussed.
Product Description
Articaine hydrochloride, European Pharmacopoeia (EP) Reference Standard