Merck
  • Weight homeostasis & its modulators in hyperthyroidism before & after treatment with carbimazole.
Weight homeostasis & its modulators in hyperthyroidism before & after treatment with carbimazole.
The Indian journal of medical research (2012-09-11)
Pinaki Dutta, Anil Bhansali, Rama Walia, Niranjan Khandelwal, Sambit Das, Shariq Rashid Masoodi
ABSTRACT
Hyperthyroidism is associated with increased food intake, energy expenditure and altered body composition. This study was aimed to evaluate the role of adipocytokines in weight homeostasis in patients with hyperthyroidism. Patients (n=27, 11men) with hyperthyroidism (20 Graves' disease, 7 toxic multinodular goiter) with mean age of 31.3±4.2 yr and 28 healthy age and body mass index (BMI) matched controls were studied. They underwent assessment of lean body mass (LBM) and total body fat (TBF) by dual energy X-ray absorptiometer (DXA) and blood sample was taken in the fasting state for measurement of leptin, adiponectin, ghrelin, insulin, glucose and lipids. Patients were re-evaluated after 3 months of treatment as by that time all of them achieved euthyroid state with carbimazole therapy. The LBM was higher (P<0.001) in healthy controls as compared to hyperthyroid patients even after adjustment for body weight (BW), whereas total body fat was comparable between the two groups. Serum leptin levels were higher in patients with hyperthyroidism than controls (22.3±3.7 and 4.1±0.34 ng/ml, P<0.001), whereas adiponectin levels were comparable. Plasma acylated ghrelin was higher in patients than in controls (209.8±13.3 vs 106.2±8.2 pg/ml, P<0.05). Achievement of euthyroidism was associated with significant weight gain (P<0.001) and significant increase in lean body mass (P<0.001). The total body fat also increased but insignificantly from 18.4±1.8 to 19.9±1.8 kg. There was significant decrease (P<0.05) in serum leptin and acylated ghrelin but adiponectin levels remained unaltered after treatment. Serum leptin positively correlated with TBF and this correlation persisted even after adjustment for BW, BMI, gender and age (r=0.62, P=0.001). However, serum leptin and acylated ghrelin did not correlate with the presence or absence of hyperphagia. Patients with hyperthyroidism predominantly had decreased lean body mass which increased after achievement of euthyroidism with carbimazole. The hyperphagia and the alterations in weight homeostasis associated with hyperthyroidism were independent of circulating leptin and ghrelin levels.
Product Description
Carbimazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbimazole, ≥98% (HPLC)