Merck
  • A novel DAX1/NR0B1 mutation in a patient with adrenal hypoplasia congenita and hypogonadotropic hypogonadism.
A novel DAX1/NR0B1 mutation in a patient with adrenal hypoplasia congenita and hypogonadotropic hypogonadism.
Arquivos brasileiros de endocrinologia e metabologia (2013-01-09)
Claudilene Battistin, Hamilton Cabral de Menezes Filho, Sorahia Domenice, Mirian Yumie Nishi, Thais Della Manna, Hilton Kuperman, Leandra Steinmetz, Vaê Dichtchekenian, Nuvarte Setian, Durval Damiani
ABSTRACT
We report a case of adrenal hypoplasia congenita (AHC) and hypogonadotropic hypogonadism (HH) due to a novel DAX1 mutation. A 19-month-old boy with hyperpigmentation and failure to thrive came to our service for investigation. Three brothers of the patient had died due to adrenal failure, and a maternal cousin had adrenal insufficiency. Adrenoleukodystrophy was excluded. MRI showed normal pituitary and hypothalamus. Plasma hormone evaluation revealed high ACTH (up to 2,790 pg/mL), and low levels of androstenedione, DHEA-S, 11-deoxycortisol, and cortisol. At 14 years of age the patient was still prepubescent, his weight was 43.6 kg (SDS: -0.87) and his height was 161 cm (SDS: -0.36), with normal body proportions. In the GnRH test, basal and maximum values of LH and FSH were respectively 0.6/2.1 and < 1.0/< 1.0 U/L. Molecular investigation identified a novel mutation that consists of a deletion of codon 372 (AAC; asparagine) in exon 1 of DAX1. This mutation was not found in a study of 200 alleles from normal individuals. Prediction site analysis indicated that this alteration, located in the DAX1 ligand-binding domain, may damage DAX1 protein. We hypothesize that the novel (p.Asp372del) DAX1 mutation might be able to cause a disruption of DAX1 function, and is probably involved in the development of AHC and HH in this patient.
Sigma-Aldrich
Anti-NR0B1 antibody produced in rabbit, IgG fraction of antiserum
Pricing and availability is not currently available.