Merck
cis-1,5-Diaminocyclooctane: the most basic gaseous primary amine?
Chemical communications (Cambridge, England) (2003-08-26)
John C Poutsma, Erica J Andriole, Tristan Sissung, Thomas Hellman Morton
ABSTRACT
The gas phase basicity of the title compound has been determined to be greater than that of putrescine, making it the most basic primary diamine measured to date.
Sigma-Aldrich
cis-1,5-Cyclooctanediol, 98%