Merck
  • [Effect of compound panax notoginsenoside granules on TGF-β1/Smads signaling pathway in rats with bleomycin-induced pulmonary fibrosis].
[Effect of compound panax notoginsenoside granules on TGF-β1/Smads signaling pathway in rats with bleomycin-induced pulmonary fibrosis].
Xi bao yu fen zi mian yi xue za zhi = Chinese journal of cellular and molecular immunology (2013-09-10)
Juanjuan Wu, Chunhua Ling, Huanrong He, Gang Sun, Zhenlun Gu, Bing Ji
ABSTRACT
To investigate the intervention effects and mechanisms of the compound panax notoginsenoside granules (CPNG) on bleomycin-induced pulmonary fibrosis in rats. Sixty male Sprague-Dawley rats were randomly divided into 6 groups, including normal control group, model group, prednisolone group, and CPNG (high, middle and low-dose) groups. Each group included 10 Sprague-Dawley rats. Except the control group, pulmonary fibrosis model rats were injected with bleomycin (5 mg/kg) via trachea. On the second day of the bleomycin injection, the rats in each treatment group were given prednisolone (3.33 mg/kg) and corresponding doses of CPNG (100, 50, 25 mg/kg) by nasogastric feeding daily; the normal control group and the model group were given the equal volume distilled water daily. Twenty-eight days later, all rats were executed and the lung tissues were subjected to the histopathological examination via HE and Masson staining. The hydroxyproline (HYP) in lung tissues was assessed by alkaline hydrolysis. The expressions of TGF-β1, Smad2, and Smad7 proteins were analyzed by Western blotting. Compared with the model group, the histopathological results revealed that CPNG dramatically decreased the severity of lung inflammation and fibrosis in rats (P<0.01). Compared with the control group, the HYP in lung tissues was reduced in all of the CPNG groups (P<0.01). Moreover, CPNG inhibited the expressions of TGF-β1 or Smad2 proteins (P<0.01), but enhanced the expression of Smad7 protein in the rat lung tissues (P<0.01). CPNG has the prevention and treatment effect on the bleomycin-induced pulmonary fibrosis in rats, and it might be attributed to the regulation on the TGF-β1/Smads signaling pathway.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg