Merck
  • Physicochemical and functional properties of gelatins extracted from turkey and chicken heads.
Physicochemical and functional properties of gelatins extracted from turkey and chicken heads.
Poultry science (2013-08-21)
L Du, Z Khiari, Z Pietrasik, M Betti
ABSTRACT
Gelatins were prepared from chicken and turkey heads in a series of batch extractions at 2 different temperatures (50 and 60°C), and their composition and functional properties were evaluated. Gelatin yield from chicken and turkey heads was 52.29 and 62.76%, respectively, on a dry weight basis relative to the total collagen content in the raw materials. The gel strength of turkey gelatins varied from 332.7 to 368.4 g, which was significantly (P < 0.05) higher than that of chicken gelatins. Both chicken and turkey head gelatins had high solubility at acidic and alkaline pH values. However, turkey head gelatins showed better emulsifying and foaming properties compared with chicken gelatins.
Product Description
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture