Merck
  • Determination of benzothiazole and benzotriazole derivates in tire and clothing textile samples by high performance liquid chromatography-electrospray ionization tandem mass spectrometry.
Determination of benzothiazole and benzotriazole derivates in tire and clothing textile samples by high performance liquid chromatography-electrospray ionization tandem mass spectrometry.
Journal of chromatography. A (2013-08-10)
Rozanna Avagyan, Ioannis Sadiktsis, Gunnar Thorsén, Conny Östman, Roger Westerholm
ABSTRACT
A high performance liquid chromatography-tandem mass spectrometry method utilizing electrospray ionization in positive and negative mode has been developed for the separation and detection of benzothiazole and benzotriazole derivates. Ultra-sonication assisted solvent extraction of these compounds has also been developed and the overall method demonstrated on a selected clothing textile and an automobile tire sample. Matrix effects and extraction recoveries, as well as linearity and limits of detection have been evaluated. The calibration curves spanned over more than two orders of magnitude with coefficients of correlation R(2)>0.99 and the limits of detection and the limits of quantification were in the range 1.7-58pg injected and 18-140pg/g, respectively. The extraction recoveries ranged between 69% and 102% and the matrix effects between 75% and 101%. Benzothiazole and benzotriazole derivates were determined in the textile sample and benzothiazole derivatives determined in the tire sample with good analytical performance.
Sigma-Aldrich
Benzothiazole, ≥96%, FG
Sigma-Aldrich
Benzothiazole, 96%