Merck
  • Development of abdominal aortic aneurysm is decreased in mice with plasma phospholipid transfer protein deficiency.
Development of abdominal aortic aneurysm is decreased in mice with plasma phospholipid transfer protein deficiency.
The American journal of pathology (2013-07-09)
Valérie Deckert, Benjamin Kretz, Ahmed Habbout, Kawtar Raghay, Jérôme Labbé, Nicolas Abello, Catherine Desrumaux, Thomas Gautier, Stéphanie Lemaire-Ewing, Guillaume Maquart, Naig Le Guern, David Masson, Eric Steinmetz, Laurent Lagrost
ABSTRACT
Plasma phospholipid transfer protein (PLTP) increases the circulating levels of proatherogenic lipoproteins, accelerates blood coagulation, and modulates inflammation. The role of PLTP in the development of abdominal aortic aneurysm (AAA) was investigated by using either a combination of mechanical and elastase injury at one site of mouse aorta (elastase model) or continuous infusion of angiotensin II in hyperlipidemic ApoE-knockout mice (Ang II model). With the elastase model, complete PLTP deficiency was associated with a significantly lower incidence and a lesser degree of AAA expansion. With the Ang II model, findings were consistent with those in the elastase model, with a lower severity grade in PLTP-deficient mice, an intermediate phenotype in PLTP-deficient heterozygotes, and a blunted effect of the PLTP-deficient trait when restricted to bone marrow-derived immune cells. The protective effect of whole-body PLTP deficiency in AAA was illustrated further by a lesser degree of adventitia expansion, reduced elastin degradation, fewer recruited macrophages, and less smooth muscle cell depletion in PLTP-deficient than in wild-type mice, as evident from comparative microscopic analysis of aorta sections. Finally, cumulative evidence supports the association of PLTP deficiency with reduced expression and activity levels of matrix metalloproteinases, known to degrade elastin and collagen. We conclude that PLTP can play a significant role in the pathophysiology of AAA.
Sigma-Aldrich
Elastin, soluble from bovine neck ligament, salt-free, lyophilized powder
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
Elastase from porcine pancreas, Type IV, Protein 50-90 %, lyophilized powder, ≥4.0 units/mg protein (biuret)
Sigma-Aldrich
Elastase from porcine pancreas, Type III, lyophilized powder, Protein 55-85 %, ≥4.0 units/mg protein
Sigma-Aldrich
Elastase from porcine pancreas, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Elastase from porcine pancreas, Type I, ≥4.0 units/mg protein