Merck
  • BINAP/1,4-diamine-ruthenium(II) complexes for efficient asymmetric hydrogenation of 1-tetralones and analogues.
BINAP/1,4-diamine-ruthenium(II) complexes for efficient asymmetric hydrogenation of 1-tetralones and analogues.
Organic letters (2004-07-30)
Takeshi Ohkuma, Tomonori Hattori, Hirohito Ooka, Tsutomu Inoue, Ryoji Noyori
ABSTRACT
A combined system of a RuCl(2)(binap)(1,4-diamine) complex and t-C(4)H(9)OK in i-C(3)H(7)OH catalyzes enantioselective hydrogenation of various 1-tetralone derivatives and some methylated 2-cyclohexenones. Hydrogenation of 2-methyl-1-tetralone under dynamic kinetic resolution gives the cis alcohol with high ee. [reaction: see text]
Sigma-Aldrich
2-Methyl-1-tetralone, 98%