Merck
  • Formation of benzynes from 2,6-dihaloaryllithiums: mechanistic basis of the regioselectivity.
Formation of benzynes from 2,6-dihaloaryllithiums: mechanistic basis of the regioselectivity.
Journal of the American Chemical Society (2004-11-13)
Antonio Ramírez, John Candler, Crystal G Bashore, Michael C Wirtz, Jotham W Coe, David B Collum
ABSTRACT
The key elimination step for the formation of 3-chloro- and 3-fluorobenzyne from 2-chloro-6-fluorophenyllithium displays a pronounced solvent-dependent regioselectivity. 6Li and 13C NMR spectroscopic studies on 2-chloro-6-fluorophenyllithium reveal a single monomeric aryllithium, suggested by DFT computational studies to be a trisolvate. Rate studies indicate that the elimination of LiCl and LiF proceeds via trisolvated and disolvated monomers, respectively.
Sigma-Aldrich
1-Chloro-3-fluorobenzene, 99%