Merck
  • Rab14 and its exchange factor FAM116 link endocytic recycling and adherens junction stability in migrating cells.
Rab14 and its exchange factor FAM116 link endocytic recycling and adherens junction stability in migrating cells.
Developmental cell (2012-05-19)
Andrea Linford, Shin-ichiro Yoshimura, Ricardo Nunes Bastos, Lars Langemeyer, Andreas Gerondopoulos, Daniel J Rigden, Francis A Barr
ABSTRACT
Rab GTPases define the vesicle trafficking pathways underpinning cell polarization and migration. Here, we find that Rab4, Rab11, and Rab14 and the candidate Rab GDP-GTP exchange factors (GEFs) FAM116A and AVL9 are required for cell migration. Rab14 and its GEF FAM116A localize to and act on an intermediate compartment of the transferrin-recycling pathway prior to Rab11 and after Rab5 and Rab4. This Rab14 intermediate recycling compartment has specific functions in migrating cells discrete from early and recycling endosomes. Rab14-depleted cells show increased N-cadherin levels at junctional complexes and cannot resolve cell-cell junctions. This is due to decreased shedding of cell-surface N-cadherin by the ADAM family protease ADAM10/Kuzbanian. In FAM116A- and Rab14-depleted cells, ADAM10 accumulates in a transferrin-positive endocytic compartment, and the cell-surface level of ADAM10 is correspondingly reduced. FAM116 and Rab14 therefore define an endocytic recycling pathway needed for ADAM protease trafficking and regulation of cell-cell junctions.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone MB1.2, clone MB1.2, Chemicon®, from rat
Sigma-Aldrich
4-Methylumbelliferyl β-D-lactopyranoside, β-galactosidase substrate